siRNA Details
Gene Symbol: Tmprss5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030709.2 | NP_109634.2 | 9 | 953 | ||
GenBank mRNA | AB016229.1 | - | 9 | 951 | ||
AB016230.1 | - | 9 | 953 | |||
AB016423.1 | - | 5 | 568 | |||
AB041037.1 | - | 9 | 910 | |||
AK029714.1 | - | 10 | 1800 | |||
BC132151.1 | - | 8 | 899 | 40130428 | ||
BC145027.1 | - | 8 | 881 | 9053575 |

More Information
Other Species Targeted

Gene Ontology Categories:
