siRNA Details
Gene Symbol: Fgf16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030614.2 | NP_085117.2 | 3 | 612 | ||
GenBank mRNA | AB049219.1 | - | 3 | 548 | ||
AF292104.1 | - | 3 | 548 | |||
BC132641.1 | - | 3 | 612 | 40130918 | ||
BC137993.1 | - | 3 | 612 | 8861012 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_021867, BC085738, AB002561.
Gene Ontology Categories:
