siRNA Details
Gene Symbol: Dtx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030714.2 | NP_109639.1 | 5 | 1230 | ||
GenBank mRNA | AB015425.1 | - | 4 | 1212 | ||
AK148444.1 | - | 5 | 1249 | |||
AK180250.1 | - | 1 | 30 | |||
AK180771.1 | - | 1 | 30 | |||
AK181396.1 | - | 1 | 30 | |||
AK189235.1 | - | 1 | 30 | |||
AK195108.1 | - | 1 | 30 | |||
AK207039.1 | - | 1 | 30 | |||
AK209833.1 | - | 1 | 30 | |||
AK218545.1 | - | 1 | 30 | |||
AK220046.1 | - | 1 | 30 | |||
BC099687.1 | - | 5 | 1230 | 6311735 | ||
BC138304.1 | - | 4 | 1109 | 8861323 | ||
BC157899.1 | - | 4 | 1111 | 9088091 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
