siRNA Details
Gene Symbol: Dtx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030714.2 | NP_109639.1 | 4 | 1009 | ||
GenBank mRNA | AB015425.1 | - | 3 | 991 | ||
AK148444.1 | - | 4 | 1028 | |||
AK199559.1 | - | 1 | 90 | |||
BC099687.1 | - | 4 | 1009 | 6311735 | ||
BC138304.1 | - | 3 | 888 | 8861323 | ||
BC157899.1 | - | 3 | 890 | 9088091 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
