siRNA Details
Gene Symbol: Gatsl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030719.3 | NP_109644.2 | 8 | 1174 | ||
GenBank mRNA | AJ296173.1 | - | 8 | 1214 | ||
AK039122.1 | - | 8 | 1200 | |||
AK052217.1 | - | 8 | 1159 | |||
BC094613.1 | - | 8 | 1174 | 6856325 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
