siRNA Details
Gene Symbol: Pum2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160219.1 | NP_001153691.1 | 20, 21 | 3317 | ||
NM_001160220.1 | NP_001153692.1 | 20, 21 | 3311 | |||
NM_001160221.1 | NP_001153693.1 | 20, 21 | 3126 | |||
NM_001160222.1 | NP_001153694.1 | 19, 20 | 2889 | |||
NM_030723.2 | NP_109648.2 | 20, 21 | 3241 | |||
GenBank mRNA | AF315590.1 | - | 20, 21 | 3257 | ||
AK048119.1 | - | 14, 15 | 2180 | |||
AK122225.1 | - | 21, 22 | 3159 | |||
AK134185.1 | - | 20, 21 | 3125 | |||
AK141682.1 | - | 20, 21 | 3173 | |||
AK147617.1 | - | 21, 22 | 3259 | |||
AK161357.1 | - | 9, 10 | 1482 | |||
AK163964.1 | - | 19, 20 | 2864 | |||
AY027917.1 | - | 20, 21 | 3157 | |||
BC013765.1 | - | 4, 5 | 492 | |||
BC041773.1 | - | 18, 19 | 2863 | 3986207 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
