siRNA Details
Gene Symbol: Pum2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160219.1 | NP_001153691.1 | 17 | 2838 | ||
NM_001160220.1 | NP_001153692.1 | 17 | 2832 | |||
NM_001160221.1 | NP_001153693.1 | 17 | 2647 | |||
NM_001160222.1 | NP_001153694.1 | 16 | 2410 | |||
NM_030723.2 | NP_109648.2 | 17 | 2762 | |||
GenBank mRNA | AF315590.1 | - | 17 | 2778 | ||
AK048119.1 | - | 11 | 1701 | |||
AK122225.1 | - | 18 | 2680 | |||
AK134185.1 | - | 17 | 2646 | |||
AK141682.1 | - | 17 | 2694 | |||
AK147617.1 | - | 18 | 2780 | |||
AK161357.1 | - | 6 | 1003 | |||
AK163964.1 | - | 16 | 2385 | |||
AY027917.1 | - | 17 | 2678 | |||
BC013765.1 | - | 1 | 13 | |||
BC041773.1 | - | 15 | 2384 | 3986207 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
