siRNA Details
Gene Symbol: Rad54l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030730.2 | NP_109655.2 | 16 | 2833 | ||
GenBank mRNA | AJ132389.1 | - | 16 | 2734 | ||
AK083903.1 | - | 16 | 2814 | |||
AK143506.1 | - | 15 | 2449 | |||
AK155151.1 | - | 16 | 2790 | |||
BC133714.1 | - | 16 | 2682 | 40143565 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001134520, XM_008766604.
Gene Ontology Categories:
