siRNA Details
Gene Symbol: Ift122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167763.1 | NP_001161235.1 | 16 | 1982 | ||
NM_031177.4 | NP_112454.2 | 16 | 1982 | |||
GenBank mRNA | AK031308.1 | - | 16 | 1870 | ||
AK044456.1 | - | 16 | 1982 | |||
AK077104.1 | - | 3 | 445 | |||
AK131115.1 | - | 16 | 1909 | |||
AK167330.1 | - | 16 | 1937 | |||
AK167412.1 | - | 16 | 1937 | |||
BC038826.1 | - | 1 | 265 | |||
BC060621.1 | - | 16 | 2069 | |||
BC066083.1 | - | 16 | 1901 | 30355922 | ||
BC067415.1 | - | 16 | 1898 | 30063124 |
