siRNA Details
Gene Symbol: Klb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031180.2 | NP_112457.1 | 2 | 1218 | ||
GenBank mRNA | AF165170.1 | - | 2 | 1218 | ||
AF178429.1 | - | 2 | 1213 | |||
AK133823.1 | - | 2 | 1245 | |||
BC138008.1 | - | 2 | 1307 | 8861027 | ||
BC138010.1 | - | 2 | 1307 | 8861029 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
