siRNA Details
Gene Symbol: Glis2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031184.3 | NP_112461.2 | 5 | 874 | ||
GenBank mRNA | AF249340.1 | - | 4 | 656 | ||
AF325913.1 | - | 4 | 626 | |||
BC016124.1 | - | 3 | 699 | |||
BC021517.1 | - | 5 | 703 | 5352352 | ||
CN703218.1 | - | 4 | 345 | |||
CX563691.1 | - | 4 | 551 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
