siRNA Details
Gene Symbol: Gimap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031247.3 | NP_112537.2 | 5 | 1648 | ||
GenBank mRNA | AB164418.1 | - | 5 | 1648 | ||
AF337052.1 | - | 5 | 1655 | |||
AK008376.1 | - | 1 | 331 | |||
BC094914.1 | - | 5 | 1628 | 30074259 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
