siRNA Details
Gene Symbol: Pramel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031377.2 | NP_113554.1 | 4 | 1352 | ||
GenBank mRNA | AF285578.1 | - | 4 | 1387 | ||
BC131995.1 | - | 4 | 1353 | 40130272 | ||
BC132399.1 | - | 4 | 1353 | 40130676 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
