siRNA Details
Gene Symbol: Lin28a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145833.1 | NP_665832.1 | 3, 4 | 489 | ||
GenBank mRNA | AB221669.1 | - | 3, 4 | 414 | ||
AF521097.1 | - | 3, 4 | 489 | |||
BC068304.1 | - | 3, 4 | 446 | 30432006 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001109269.
Gene Ontology Categories:
