siRNA Details
Gene Symbol: Akr1c6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030611.3 | NP_085114.1 | 8 | 956 | ||
GenBank mRNA | AK149458.1 | - | 8 | 957 | ||
BC056643.1 | - | 8 | 938 | 5098276 | ||
D45850.1 | - | 8 | 908 |
Gene Symbol: Akr1c20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB097220.1 | - | 8 | 915 | ||
BC021607.1 | - | 8 | 923 | 5098938 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
