siRNA Details
Gene Symbol: Dbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031403.3 | NP_113580.2 | 4 | 572 | ||
GenBank mRNA | AK050090.1 | - | 4 | 573 | ||
AK090351.1 | - | 4 | 548 | |||
AK139631.1 | - | 4 | 554 | |||
BC006661.1 | - | 4 | 567 | 3501390 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
