siRNA Details
Gene Symbol: Cep41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ278891.1 | - | 5 | 271 | ||
AK006059.1 | - | 5 | 259 | |||
AK132495.1 | - | 5 | 260 | |||
AK161461.1 | - | 5 | 245 | |||
AK166974.1 | - | 5 | 252 | |||
AK170213.1 | - | 5 | 284 | |||
BC031892.1 | - | 5 | 241 | 5365552 | ||
BY736279.1 | - | 5 | 252 |
