siRNA Details
Gene Symbol: Trps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032000.2 | NP_114389.2 | 2 | 1174 | ||
GenBank mRNA | AF346836.1 | - | 3 | 1099 | ||
AK142355.1 | - | 2 | 844 | |||
BC037058.1 | - | 2 | 852 | 4951294 | ||
BC049857.1 | - | 4 | 740 | 4460789 | ||
BC099962.1 | - | 2 | 1139 | 30091008 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001134837, XM_008765496, XM_008765498, XM_008765497, XM_008765499.
Gene Ontology Categories:
