siRNA Details
Gene Symbol: Trps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032000.2 | NP_114389.2 | 6 | 3596 | ||
GenBank mRNA | AF346836.1 | - | 7 | 3521 | ||
AK142355.1 | - | 6 | 3266 | |||
BC037058.1 | - | 8 | 2707 | 4951294 | ||
BC049857.1 | - | 9 | 2562 | 4460789 | ||
BC083110.1 | - | 5 | 2354 | |||
BC099962.1 | - | 6 | 3561 | 30091008 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
