siRNA Details
Gene Symbol: Nrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032002.2 | NP_114391.1 | 7 | 692 | ||
GenBank mRNA | AF083067.2 | - | 6 | 570 | ||
AK041077.1 | - | 8 | 686 | |||
AK080058.1 | - | 7 | 644 | |||
AK080161.1 | - | 9 | 786 | |||
BC034839.1 | - | 10 | 985 | 3372942 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
