siRNA Details
Gene Symbol: Nrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032002.2 | NP_114391.1 | 3, 4 | 286 | ||
GenBank mRNA | AF083067.2 | - | 2, 3 | 164 | ||
AK080089.1 | - | 3, 4 | 264 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
