siRNA Details
Gene Symbol: Btbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146193.2 | NP_666305.2 | 6 | 1179 | ||
GenBank mRNA | AK088345.1 | - | 6 | 1220 | ||
AK215939.1 | - | 2 | 101 | |||
AK217049.1 | - | 2 | 101 | |||
BC031192.1 | - | 5 | 657 | 5043252 | ||
BC072618.1 | - | 6 | 1179 | 6811133 | ||
BC087937.1 | - | 6 | 1212 | 6705214 |
