siRNA Details
Gene Symbol: Jam3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023277.4 | NP_075766.1 | 4 | 452 | ||
GenBank mRNA | AJ300304.1 | - | 4 | 412 | ||
AK003326.1 | - | 4 | 412 | |||
AK008187.1 | - | 4 | 445 | |||
AK013156.1 | - | 4 | 411 | |||
AK032833.1 | - | 4 | 412 | |||
AK160648.1 | - | 4 | 450 | |||
BC024357.1 | - | 4 | 424 | 4973185 |

More Information
Other Species Targeted

Gene Ontology Categories:
