siRNA Details
Gene Symbol: Setdb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163641.1 | NP_001157113.1 | 3 | 503 | ||
NM_001163642.1 | NP_001157114.1 | 3 | 465 | |||
NM_018877.3 | NP_061365.3 | 3 | 465 | |||
GenBank mRNA | AF091628.1 | - | 3 | 419 | ||
AF546078.1 | - | 3 | 419 | |||
AK132202.1 | - | 3 | 437 | |||
AK135833.1 | - | 3 | 416 | |||
AY091600.1 | - | 3 | 419 | |||
BC060173.1 | - | 3 | 459 | |||
BC079537.1 | - | 3 | 418 | 6410215 |
Gene Symbol: Gm4349 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AV576091.1 | - | 3 | 378 |
