siRNA Details
Gene Symbol: Il24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053095.2 | NP_444325.2 | 5, 6 | 648 | ||
GenBank mRNA | AF235006.1 | - | 4, 5 | 714 | ||
AF333251.1 | - | 5, 6 | 576 | |||
DQ401033.1 | - | 5, 6 | 547 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
