siRNA Details
Gene Symbol: Cml3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037842.3 | NP_001032931.1 | 3 | 1110 | ||
GenBank mRNA | AK046314.1 | - | 3 | 1111 | ||
BC014696.1 | - | 848 | 4237604 |
Gene Symbol: Gm11128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201389.1 | NP_001188318.1 | 3 | 677 | ||
GenBank mRNA | AF163316.1 | - | 3 | 676 |
Gene Symbol: Gm4477 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253910.2 | NP_001240839.2 | 545 | |||
GenBank mRNA | AB221545.1 | - | 1 | 424 | ||
BC092089.1 | - | 538 | 4975242 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
