siRNA Details
Gene Symbol: Lmod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053098.2 | NP_444328.1 | 2 | 1163 | ||
GenBank mRNA | AF237628.1 | - | 1 | 400 | ||
AK147141.1 | - | 2 | 1164 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
