siRNA Details
Gene Symbol: Lmod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053098.2 | NP_444328.1 | 2 | 942 | ||
GenBank mRNA | AF237628.1 | - | 1 | 179 | ||
AK147141.1 | - | 2 | 943 |

More Information
Other Species Targeted

Gene Ontology Categories:
