siRNA Details
Gene Symbol: Glrx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053108.4 | NP_444338.2 | 1 | 249 | ||
GenBank mRNA | AB013137.1 | - | 1 | 215 | ||
AF109314.1 | - | 1 | 225 | |||
AF276917.1 | - | 1 | 174 | |||
AK013722.1 | - | 1 | 231 | |||
AK153086.1 | - | 1 | 229 | |||
BC012642.1 | - | 1 | 226 | 4206114 | ||
BC054418.1 | - | 1 | 211 | |||
BY272278.1 | - | 1 | 268 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
