siRNA Details
Gene Symbol: Chrac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053068.3 | NP_444298.1 | 3 | 320 | ||
GenBank mRNA | AF230805.1 | - | 3 | 295 | ||
AK010812.1 | - | 3 | 321 | |||
AK134403.1 | - | 871 | ||||
BC016593.1 | - | 3 | 292 | |||
BC061043.1 | - | 3 | 319 | 6773451 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001134880, FQ209946.
Gene Ontology Categories:
