siRNA Details
Gene Symbol: Pcdhgb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033574.3 | NP_291052.3 | 1 | 162 | ||
GenBank mRNA | AK082842.1 | - | 1 | 364 | ||
AY013804.1 | - | 1 | 162 | |||
BC054741.1 | - | 1 | 329 | 5721217 |
