siRNA Details
Gene Symbol: Pcdhga8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033591.3 | NP_291069.1 | 1 | 1438 | ||
GenBank mRNA | AK220461.1 | - | 1 | 1391 | ||
AY013802.1 | - | 1 | 1251 | |||
BC054555.1 | - | 1 | 1399 | 5698395 | ||
BC054787.1 | - | 1 | 1438 | 6825772 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
