siRNA Details
Gene Symbol: Cpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030703.2 | NP_109628.1 | 6 | 1124 | ||
GenBank mRNA | AB021969.1 | - | 6 | 1101 | ||
AF326477.1 | - | 6 | 1124 | |||
AK033629.1 | - | 2 | 263 | |||
BC014692.1 | - | 6 | 1092 | 4211169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
