siRNA Details
Gene Symbol: Rnase2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053113.2 | NP_444343.2 | 2 | 418 | ||
GenBank mRNA | BC027557.1 | - | 2 | 418 | 1445853 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
