siRNA Details
Gene Symbol: Acox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161667.1 | NP_001155139.1 | 13 | 1874 | ||
NM_053115.2 | NP_444345.2 | 13 | 1999 | |||
GenBank mRNA | AJ238492.1 | - | 13 | 1916 | ||
AK144243.1 | - | 13 | 1868 | |||
BC021339.1 | - | 13 | 1878 | 5052291 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
