siRNA Details
Gene Symbol: Mpv17l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289561.1 | NP_001276490.1 | 4 | 603 | ||
NM_001289562.1 | NP_001276491.1 | 4 | 335 | |||
NM_001289563.1 | NP_001276492.1 | 4 | 298 | |||
NM_001289565.1 | NP_001276494.1 | 5 | 352 | |||
NM_001289567.1 | NP_001276496.1 | 5 | 628 | |||
NM_033564.3 | NP_291042.2 | 4 | 574 | |||
GenBank mRNA | AF305634.1 | - | 4 | 544 | ||
AK034151.1 | - | 1 | 50 | |||
AK136449.1 | - | 4 | 336 | |||
AK144372.1 | - | 4 | 297 | |||
AY513273.1 | - | 4 | 462 | |||
BC037713.1 | - | 4 | 454 | |||
BC094450.1 | - | 4 | 541 | 4973897 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
