siRNA Details
Gene Symbol: Rnf111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033604.2 | NP_291082.1 | 12 | 2922 | ||
GenBank mRNA | AF330197.1 | - | 12 | 2918 | ||
AK036351.1 | - | 11 | 2657 | |||
AK048110.1 | - | 11 | 2688 | |||
AK137148.1 | - | 10 | 2260 | |||
AK164502.1 | - | 3 | 319 | |||
AK169999.1 | - | 11 | 3028 | |||
BC011463.1 | - | 6 | 1354 | |||
BC026485.1 | - | 3 | 283 | |||
BC054842.1 | - | 2 | 161 | |||
BC069835.1 | - | 12 | 2778 | 30094289 |
