siRNA Details
Gene Symbol: Rnf111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033604.2 | NP_291082.1 | 12 | 2919 | ||
GenBank mRNA | AF330197.1 | - | 12 | 2915 | ||
AK036351.1 | - | 11 | 2654 | |||
AK048110.1 | - | 11 | 2685 | |||
AK137148.1 | - | 10 | 2257 | |||
AK164502.1 | - | 3 | 316 | |||
AK169999.1 | - | 11 | 3025 | |||
BC011463.1 | - | 6 | 1351 | |||
BC026485.1 | - | 3 | 280 | |||
BC054842.1 | - | 2 | 158 | |||
BC069835.1 | - | 12 | 2775 | 30094289 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
