siRNA Details
Gene Symbol: Uchl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033607.1 | NP_291085.1 | 1 | 376 | ||
GenBank mRNA | AB035420.1 | - | 1 | 376 | ||
BC139440.1 | - | 1 | 367 | 8862462 | ||
BC139441.1 | - | 1 | 367 | 8862463 |
