siRNA Details
Gene Symbol: Brwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103179.1 | NP_001096649.1 | 41 | 6594 | ||
NM_145125.3 | NP_660107.2 | 41 | 6594 | |||
GenBank mRNA | AJ292467.1 | - | 41 | 6599 | ||
AJ292468.1 | - | 41 | 6599 | |||
AK162170.1 | - | 1 | 288 | |||
BC145265.1 | - | 41 | 6520 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
