siRNA Details
Gene Symbol: Brwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103179.1 | NP_001096649.1 | 28 | 3388 | ||
NM_145125.3 | NP_660107.2 | 28 | 3388 | |||
GenBank mRNA | AJ292467.1 | - | 28 | 3393 | ||
AJ292468.1 | - | 28 | 3393 | |||
BC145265.1 | - | 28 | 3314 | |||
CF162350.1 | - | 5 | 473 | |||
CV558558.1 | - | 2 | 148 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
