siRNA Details
Gene Symbol: Pcdhb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053126.1 | NP_444356.1 | 1 | 2312 | ||
GenBank mRNA | AY013770.1 | - | 1 | 2312 | ||
BC132112.1 | - | 1 | 2372 | 40130389 | ||
BC132296.1 | - | 1 | 2372 | 40130573 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
