siRNA Details
Gene Symbol: Pcdhb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053131.1 | NP_444361.1 | 1 | 679 | ||
GenBank mRNA | AK032193.1 | - | 292 | |||
AY013788.1 | - | 1 | 679 | |||
BC150716.1 | - | 1 | 761 | 9087627 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
