siRNA Details
Gene Symbol: Pcdhb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053132.3 | NP_444362.3 | 1 | 302 | ||
GenBank mRNA | AF326300.1 | - | 1 | 98 | ||
AK029340.1 | - | 1 | 281 | |||
AK034205.1 | - | 1 | 304 | |||
AK043979.1 | - | 1 | 261 | |||
AK046463.1 | - | 1 | 285 | |||
AY013789.1 | - | 1 | 98 | |||
BC094897.1 | - | 1 | 264 | |||
BY245516.1 | - | 1 | 303 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
