siRNA Details
Gene Symbol: Pcdhb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053136.3 | NP_444366.3 | 1 | 880 | ||
GenBank mRNA | AK078189.1 | - | 1 | 881 | ||
AK160803.1 | - | 1 | 849 | |||
AY013772.1 | - | 1 | 679 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
