siRNA Details
Gene Symbol: Pcdhb15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053140.3 | NP_444370.1 | 1 | 1116 | ||
GenBank mRNA | AK038663.1 | - | 1 | 1118 | ||
AY013776.1 | - | 1 | 945 | |||
BC066178.1 | - | 1 | 1122 | 30100073 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
