siRNA Details
Gene Symbol: Glp2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175681.3 | NP_783612.2 | 6 | 804 | ||
GenBank mRNA | AF166265.1 | - | 2 | 44 | ||
AY255567.1 | - | 5 | 418 | |||
AY605231.1 | - | 6 | 523 | |||
BC044746.2 | - | 6 | 804 | 5363416 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
