siRNA Details
Gene Symbol: Klra21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053151.1 | NP_444381.1 | 650 | |||
GenBank mRNA | AF288380.1 | - | 650 | |||
AY971806.1 | - | 544 | ||||
BC116399.1 | - | 618 | 40101978 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
