siRNA Details
Gene Symbol: Allc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053156.2 | NP_444386.2 | 7 | 674 | ||
GenBank mRNA | AF278712.1 | - | 7 | 682 | ||
BC108957.1 | - | 7 | 674 | 40046116 | ||
BC108958.1 | - | 7 | 674 | 40046117 |

More Information
Other Species Targeted

Gene Ontology Categories:
