siRNA Details
Gene Symbol: Bcl2l13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153516.2 | NP_705736.1 | 4 | 505 | ||
GenBank mRNA | AK031018.1 | - | 4 | 454 | ||
AK047223.1 | - | 4 | 507 | |||
AK088417.1 | - | 4 | 520 | |||
AK132774.1 | - | 4 | 514 | |||
AK143457.1 | - | 4 | 518 | |||
AK148007.1 | - | 4 | 524 | |||
AK165068.1 | - | 4 | 498 | |||
AK168833.1 | - | 4 | 495 | |||
AK171256.1 | - | 4 | 525 | |||
AK178476.1 | - | 3 | 280 | |||
AK178590.1 | - | 3 | 278 | |||
AK179659.1 | - | 3 | 277 | |||
AK186867.1 | - | 3 | 278 | |||
AK187777.1 | - | 3 | 278 | |||
BC027307.1 | - | 4 | 395 | 4014844 | ||
BC029016.1 | - | 4 | 505 | 5358785 |
